5-Amino-3-(4-chlorophenyl)-1,2,4-thiadiazole, 96%, Thermo Scientific Chemicals
5-Amino-3-(4-chlorophenyl)-1,2,4-thiadiazole, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Amino-3-(4-chlorophenyl)-1,2,4-thiadiazole, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Catalog number H33467.03
also known as H33467-03
Price (USD)/ Each
210.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
210.00
Add to cart
5-Amino-3-(4-chlorophenyl)-1,2,4-thiadiazole, 96%, Thermo Scientific Chemicals
Catalog numberH33467.03
Price (USD)/ Each
210.00
-
Add to cart
Chemical Identifiers
CAS19922-07-7
IUPAC Name3-(4-chlorophenyl)-1,2,4-thiadiazol-5-amine
Molecular FormulaC8H6ClN3S
InChI KeyIUSHKZWNRIGLGO-UHFFFAOYSA-N
SMILESNC1=NC(=NS1)C1=CC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)Pale cream
Assay (GC)≥95.0%
Melting Point (clear melt)175.0-184.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only