4-(3-Trifluoromethyl-2-pyridyloxy)benzenesulfonyl chloride, 97%, Thermo Scientific Chemicals
4-(3-Trifluoromethyl-2-pyridyloxy)benzenesulfonyl chloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-(3-Trifluoromethyl-2-pyridyloxy)benzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 mg
Catalog number H33473.06
also known as H33473-06
Price (USD)/ Each
384.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
384.00
Add to cart
4-(3-Trifluoromethyl-2-pyridyloxy)benzenesulfonyl chloride, 97%, Thermo Scientific Chemicals
Catalog numberH33473.06
Price (USD)/ Each
384.00
-
Add to cart
Chemical Identifiers
CAS874839-13-1
IUPAC Name4-{[3-(trifluoromethyl)pyridin-2-yl]oxy}benzene-1-sulfonyl chloride
Molecular FormulaC12H7ClF3NO3S
InChI KeyFDAYBQVWHDJJOL-UHFFFAOYSA-N
SMILESFC(F)(F)C1=C(OC2=CC=C(C=C2)S(Cl)(=O)=O)N=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Melting Point (clear melt)62.5-71.5?C
Assay (GC)≥96.0%
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only