Thermo Scientific Chemicals

Oleyl alcohol, 99+%, Thermo Scientific Chemicals

Catalog number: H33475.14
25 g, Each
Thermo Scientific Chemicals

Oleyl alcohol, 99+%, Thermo Scientific Chemicals

Catalog number: H33475.14
25 g, Each
Quantity
Catalog number: H33475.14
also known as H33475-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
143-28-2
IUPAC Name
(9Z)-octadec-9-en-1-ol
Molecular Formula
C18H36O
InChI Key
ALSTYHKOOCGGFT-KTKRTIGZSA-N
SMILES
CCCCCCCC\C=C/CCCCCCCCO
Appearance (Color)
Clear colorless
Form
Liquid
Refractive Index
1.4600-1.4620 @ 20?C
Assay (GC)
≥99.0%

Description

Oleyl alcohol is used in softening and lubrication of textile fabrics, in the production of carbon paper, stencil paper, and printing ink. It finds application in cosmetic products viz skin creams, lotions as a thickner, and in hair conditioners and hair coating shampoos. It is utilized as an antifoaming agent and cutting lubricant, as the precursor for the preparation of its sulfuric ester derivatives, which are used in detergents and wetting agents. It plays a vital role in various formulations for drug delivery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Oleyl alcohol is used in softening and lubrication of textile fabrics, in the production of carbon paper, stencil paper, and printing ink. It finds application in cosmetic products viz skin creams, lotions as a thickner, and in hair conditioners and hair coating shampoos. It is utilized as an antifoaming agent and cutting lubricant, as the precursor for the preparation of its sulfuric ester derivatives, which are used in detergents and wetting agents. It plays a vital role in various formulations for drug delivery.

Solubility
Miscible with alcohol and ether. Slightly miscible with carbon tetrachloride. Immiscible with water.

Notes
Incompatible with oxidizing agents and strong acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text