2-Aminopyridine-6-methanol, 97%, Thermo Scientific Chemicals
2-Aminopyridine-6-methanol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Aminopyridine-6-methanol, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H33485.03
also known as H33485-03
Price (USD)/ Each
210.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
210.00
Add to cart
2-Aminopyridine-6-methanol, 97%, Thermo Scientific Chemicals
Catalog numberH33485.03
Price (USD)/ Each
210.00
-
Add to cart
Chemical Identifiers
CAS79651-64-2
IUPAC Name(6-aminopyridin-2-yl)methanol
Molecular FormulaC6H8N2O
InChI KeyYGMPMXTWKROXPP-UHFFFAOYSA-N
SMILESNC1=CC=CC(CO)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale brown
Melting Point (clear melt)85.0-95.0?C
Assay (GC)≥96.0%
FormPowder
4-Aminopyridine (4-AP), a well-known potassium channel blocker, has been shown to enhance conduction in injured and demyelinated axons. Aminopyridine has a narrow therapeutic range in clinical application. In the current study, we investigated whether AP-MeOH can enhance impulse conduction in an animal model of MS.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Slightly soluble.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Jinfeng Zhao,; Peng Song,; Yanling Cui,; Xuemei Liu,; Shaowu Sun,; Siyao Hou,; Fengcai Ma. Effects of hydrogen bond on 2-aminopyridine and its derivatives complexes in methanol solvent. Spectrochimica Acta Part A: Molecular and Biomolecular Spectroscopy. 2015, 131 282-287.
  2. Gary Leung,; Wenjing Sun,; Sarah Brookes,; Daniel Smith,; Riyi Shi. Potassium channel blocker, 4-aminopyridine-3-methanol, restores axonal conduction in spinal cord of an animal model of multiple sclerosis. Experimental Neurology. year of publication , 227 (1), 232-235.