Thermo Scientific Chemicals

2-Aminopyridine-6-methanol, 97%, Thermo Scientific Chemicals

Catalog number: H33485.06
5 g, Each
Thermo Scientific Chemicals

2-Aminopyridine-6-methanol, 97%, Thermo Scientific Chemicals

Catalog number: H33485.06
5 g, Each
Quantity
Catalog number: H33485.06
also known as H33485-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
79651-64-2
IUPAC Name
(6-aminopyridin-2-yl)methanol
Molecular Formula
C6H8N2O
InChI Key
YGMPMXTWKROXPP-UHFFFAOYSA-N
SMILES
NC1=CC=CC(CO)=N1
Appearance (Color)
Pale brown
Melting Point (clear melt)
85.0-95.0?C
Assay (GC)
≥96.0%
Form
Powder

Description

4-Aminopyridine (4-AP), a well-known potassium channel blocker, has been shown to enhance conduction in injured and demyelinated axons. Aminopyridine has a narrow therapeutic range in clinical application. In the current study, we investigated whether AP-MeOH can enhance impulse conduction in an animal model of MS.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Slightly soluble.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text