5-Bromo-3-iodo-2-hydroxypyridine, 97%, Thermo Scientific Chemicals
5-Bromo-3-iodo-2-hydroxypyridine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Bromo-3-iodo-2-hydroxypyridine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number H33495.06
also known as H33495-06
Price (USD)/ Each
183.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
183.00
Add to cart
5-Bromo-3-iodo-2-hydroxypyridine, 97%, Thermo Scientific Chemicals
Catalog numberH33495.06
Price (USD)/ Each
183.00
-
Add to cart
Chemical Identifiers
CAS381233-75-6
IUPAC Name5-bromo-3-iodo-1,2-dihydropyridin-2-one
Molecular FormulaC5H3BrINO
InChI KeyWAUPPDAXWKLNNU-UHFFFAOYSA-N
SMILESBrC1=CNC(=O)C(I)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to yellow
FormCrystals or powder or crystalline powder
Assay (Silylated GC)≥96.0%
Melting Point (clear melt)227.0-233.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only