4-Fluorobenzoic anhydride, 97%, Thermo Scientific Chemicals
4-Fluorobenzoic anhydride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Fluorobenzoic anhydride, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H33527.06
also known as H33527-06
Price (USD)/ Each
292.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
292.00
Add to cart
4-Fluorobenzoic anhydride, 97%, Thermo Scientific Chemicals
Catalog numberH33527.06
Price (USD)/ Each
292.00
-
Add to cart
Chemical Identifiers
CAS25569-77-1
IUPAC Name4-fluorobenzoyl 4-fluorobenzoate
Molecular FormulaC14H8F2O3
InChI KeyBBLXFRIGTQYGOT-UHFFFAOYSA-N
SMILESFC1=CC=C(C=C1)C(=O)OC(=O)C1=CC=C(F)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)109.5-118.5?C
Appearance (Color)White
Assay (GC)≥96.0%
FormCrystals or powder or crystalline powder
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. C. Elisabet Tranberg.; Andrea Zickgraf.; Brian N. Giunta.; Henning Luetjens.; Heidi Figler.; Lauren J. Murphree.; Ruediger Falke.; Holger Fleischer.; Joel Linden.; Peter J. Scammells.; Ray. A. Olsson. 2-Amino-3-aroyl-4,5-alkylthiophenes:  Agonist Allosteric Enhancers at Human A1 Adenosine Receptors.J. Med. Chem. 2002, 45 (2),382-389 .
  2. Robert H. Dodd.; Catherine Ouannes.; Malka Robert-Gero.; Pierre Potier. Hybrid molecules: growth inhibition of Leishmania donovani promastigotes by thiosemicarbazones of 3-carboxy-.beta.-carbolines.J. Med. Chem. 1989, 32 (6),1272-1276 .