Thermo Scientific Chemicals

3-Amino-5-phenyl-1,2,4-thiadiazole, 96%, Thermo Scientific Chemicals

Catalog number: H33552.MD
250 mg, Each
Thermo Scientific Chemicals

3-Amino-5-phenyl-1,2,4-thiadiazole, 96%, Thermo Scientific Chemicals

Catalog number: H33552.MD
250 mg, Each
Quantity
Catalog number: H33552.MD
also known as H33552-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
27182-54-3
IUPAC Name
5-phenyl-1,2,4-thiadiazol-3-amine
Molecular Formula
C8H7N3S
InChI Key
WOZDQVLZCXNJPY-UHFFFAOYSA-N
SMILES
NC1=NSC(=N1)C1=CC=CC=C1
Appearance (Color)
White
Melting Point (clear melt)
134.0-143.0?C
Form
Crystals or powder or crystalline powder
Assay (GC)
≥95.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text