3-(2,5-Dimethoxyphenyl)propionic acid, 96%, Thermo Scientific Chemicals
3-(2,5-Dimethoxyphenyl)propionic acid, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-(2,5-Dimethoxyphenyl)propionic acid, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number H33566.06
also known as H33566-06
Price (USD)/ Each
90.20
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
90.20
Add to cart
3-(2,5-Dimethoxyphenyl)propionic acid, 96%, Thermo Scientific Chemicals
Catalog numberH33566.06
Price (USD)/ Each
90.20
-
Add to cart
Chemical Identifiers
CAS10538-49-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Assay (Silylated GC)≥95.0%
Assay (Aqueous acid-base Titration)≥95.0 to ≤105.0%
FormPowder
Melting Point (clear melt)60.5-69.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only