2,4,6-Triphenoxy-1,3,5-triazine, 96%, Thermo Scientific Chemicals
2,4,6-Triphenoxy-1,3,5-triazine, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,4,6-Triphenoxy-1,3,5-triazine, 96%, Thermo Scientific Chemicals

Have Questions?
Quantity:
50 g
Catalog number H33572.18
also known as H33572-18
Price (USD)/ Each
149.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
149.00
Add to cart
2,4,6-Triphenoxy-1,3,5-triazine, 96%, Thermo Scientific Chemicals
Catalog numberH33572.18
Price (USD)/ Each
149.00
-
Add to cart
Chemical Identifiers
CAS1919-48-8
IUPAC Nametriphenoxy-1,3,5-triazine
Molecular FormulaC21H15N3O3
InChI KeyIYDYVVVAQKFGBS-UHFFFAOYSA-N
SMILESO(C1=CC=CC=C1)C1=NC(OC2=CC=CC=C2)=NC(OC2=CC=CC=C2)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)232.0-238.0?C
Appearance (Color)White
Assay (HPLC)≥95.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only