2,3-Dihydrobenzo[b]furan-5-sulfonyl chloride, 97%, Thermo Scientific Chemicals
2,3-Dihydrobenzo[b]furan-5-sulfonyl chloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,3-Dihydrobenzo[b]furan-5-sulfonyl chloride, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
5 g
Catalog number H33579.MD
also known as H33579-MD
Price (USD)/ Each
41.80
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
41.80
Add to cart
2,3-Dihydrobenzo[b]furan-5-sulfonyl chloride, 97%, Thermo Scientific Chemicals
Catalog numberH33579.MD
Price (USD)/ Each
41.80
-
Add to cart
Chemical Identifiers
CAS115010-11-2
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Melting Point (clear melt)79.5-88.5?C
Appearance (Color)White to pale cream
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only