Thermo Scientific Chemicals

4-Cyclohexylbenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: H33616.09
10 g, Each
Thermo Scientific Chemicals

4-Cyclohexylbenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: H33616.09
10 g, Each
Quantity
Catalog number: H33616.09
also known as H33616-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
56354-57-5
IUPAC Name
4-cyclohexylbenzene-1-sulfonyl chloride
Molecular Formula
C12H15ClO2S
InChI Key
CREMYEDHKUWVTB-UHFFFAOYSA-N
SMILES
ClS(=O)(=O)C1=CC=C(C=C1)C1CCCCC1
Melting Point (clear melt)
49.0-58.0?C
Appearance (Color)
White to pale cream
Assay (Titration ex Chloride)
≥96.0 to ≤104.0%
Form
Crystals or powder or crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text