Assay (special)≥95.0 to ≤105.0% (based on elemental C, H, Cl)
Elemental AnalysisCl: 6.20-6.86% (Theory: 6.53%)
lambda-max (UV-Vis)520-535nm (in methanol)
Appearance (Color)Pink to red to dark red
Elemental AnalysisC: 58.84-65.03% (Theory: 61.94%)
View more