Thermo Scientific Chemicals

4'-Bromo-2,2,2-trifluoroacetophenone, 97%, Thermo Scientific Chemicals

Catalog number: H33627.06
5 g, Each
Thermo Scientific Chemicals

4'-Bromo-2,2,2-trifluoroacetophenone, 97%, Thermo Scientific Chemicals

Catalog number: H33627.06
5 g, Each
Quantity
Catalog number: H33627.06
also known as H33627-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16184-89-7
IUPAC Name
1-(4-bromophenyl)-2,2,2-trifluoroethan-1-one
Molecular Formula
C8H4BrF3O
InChI Key
IHGSAQHSAGRWNI-UHFFFAOYSA-N
SMILES
FC(F)(F)C(=O)C1=CC=C(Br)C=C1
Appearance (Color)
White to pale yellow or clear colorless to pale yellow
Form
Crystals or powder or crystalline powder or fused solid or liquid as melt
Assay (GC)
≥96.0%
Melting Point (clear melt)
26.0-32.0°C
Proton NMR
Conforms to structure

Description

4?-Bromo-2,2,2-trifluoroacetophenone may be used in the preparation of carbonyl-bridged bithiazole derivatives. Also used as a reagent to synthesize MK-5046, a selective Bombesin receptor subtype-3 agonist used to treat obesity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-Bromo-2,2,2-trifluoroacetophenone may be used in the preparation of carbonyl-bridged bithiazole derivatives. Also used as a reagent to synthesize MK-5046, a selective Bombesin receptor subtype-3 agonist used to treat obesity.

Solubility
Slightly soluble in water.

Notes
Moisture Sensitive. Store away from water/moisture. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text