Thermo Scientific Chemicals

3-Bromofuro[3,2-b]pyridine, 96%, Thermo Scientific Chemicals

Catalog number: H33652.06
5 g, Each
Thermo Scientific Chemicals

3-Bromofuro[3,2-b]pyridine, 96%, Thermo Scientific Chemicals

Catalog number: H33652.06
5 g, Each
Quantity
Catalog number: H33652.06
also known as H33652-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
92404-63-2
IUPAC Name
3-bromofuro[3,2-b]pyridine
Molecular Formula
C7H4BrNO
InChI Key
KHMAAFDMZSYVEF-UHFFFAOYSA-N
SMILES
BrC1=COC2=CC=CN=C12
Appearance (Color)
Pale cream to cream to brown
Form
Crystals or powder or crystalline powder
Assay (GC)
≥95.0%
Melting Point (clear melt)
49.0-58.0?C

Description

3-Bromofuro[3,2-b]pyridine is used as a starting material for synthesis of polyheterocycles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromofuro[3,2-b]pyridine is used as a starting material for synthesis of polyheterocycles.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text