Thermo Scientific Chemicals

3-(4-Nitrophenyl)propionic acid, 96%, Thermo Scientific Chemicals

Catalog number: H33663.09
10 g, Each
Thermo Scientific Chemicals

3-(4-Nitrophenyl)propionic acid, 96%, Thermo Scientific Chemicals

Catalog number: H33663.09
10 g, Each
Quantity
Catalog number: H33663.09
also known as H33663-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16642-79-8
Melting Point (clear melt)
161.0-170.0?C
Appearance (Color)
White to yellow
Form
Crystals or powder or crystalline powder
Assay (HPLC)
≥97.5%

Description

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed containers. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text