Thermo Scientific Chemicals

12-Bromododecylphosphonic acid, 95%, Thermo Scientific Chemicals

Catalog number: H33714.MD
250 mg, Each
Thermo Scientific Chemicals

12-Bromododecylphosphonic acid, 95%, Thermo Scientific Chemicals

Catalog number: H33714.MD
250 mg, Each
Quantity
Catalog number: H33714.MD
also known as H33714-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
202920-07-8
Appearance (Color)
White
Assay (Aqueous acid-base Titration)
≥94.0 to ≤106.0%
Melting Point (clear melt)
87.5-96.5?C
Form
Powder

Description

Employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as a intermediate for pharmaceutical.

Notes
Store in cool, dry place in tightly closed containers. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text