Thermo Scientific Chemicals

4-Methylthiazole-5-sulfonyl chloride, 95%, Thermo Scientific Chemicals

Catalog number: H33732.MD
250 mg, Each
Thermo Scientific Chemicals

4-Methylthiazole-5-sulfonyl chloride, 95%, Thermo Scientific Chemicals

Catalog number: H33732.MD
250 mg, Each
Quantity
Catalog number: H33732.MD
also known as H33732-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
953070-51-4
IUPAC Name
4-methyl-1,3-thiazole-5-sulfonyl chloride
Molecular Formula
C4H4ClNO2S2
InChI Key
AWTJUYXVSWZCIK-UHFFFAOYSA-N
SMILES
CC1=C(SC=N1)S(Cl)(=O)=O
Form
Crystals or powder or crystalline powder
Appearance (Color)
White to yellow to brown
Assay (Titration ex Chloride)
≥94.0 to ≤106.0%

Description

It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text