Thermo Scientific Chemicals

Zinc acetate, anhydrous, 99.9+%, Thermo Scientific Chemicals

Catalog number: H33734.18
50 g, Each
Thermo Scientific Chemicals

Zinc acetate, anhydrous, 99.9+%, Thermo Scientific Chemicals

Catalog number: H33734.18
50 g, Each
Quantity
Catalog number: H33734.18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
557-34-6
IUPAC Name
zinc(2+) diacetate
Molecular Formula
C4H6O4Zn
InChI Key
DJWUNCQRNNEAKC-UHFFFAOYSA-L
SMILES
[Zn++].CC([O-])=O.CC([O-])=O
Elemental Analysis
Zn: 35.34 - 35.94% (dry wt. basis)
Trace Metal
Arsenic As: < 5ppm
Trace Metal
Lead Pb: < 50ppm
Appearance (Color)
White
Trace Metal
Cadmium Cd: < 10ppm

Description

Zinc acetate anhydrous is used in the synthesis of layered Zn-arylphosphonates with potential application in sorption, ion exchange or catalysis. It is utilized in the ultrasonic preparation of zinc sulfide nanoparticles coated on silica particles. It is administered orally or parenterally as a nutritional supplement. It finds an application in the field of industries such as wood preservation, manufacturing other zinc salts, polymers, manufacture of ethylene acetate, as a dye mordant, and analytical reagent. It also acts as a plating inhibitor on primary water piping.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zinc acetate anhydrous is used in the synthesis of layered Zn-arylphosphonates with potential application in sorption, ion exchange or catalysis. It is utilized in the ultrasonic preparation of zinc sulfide nanoparticles coated on silica particles. It is administered orally or parenterally as a nutritional supplement. It finds an application in the field of industries such as wood preservation, manufacturing other zinc salts, polymers, manufacture of ethylene acetate, as a dye mordant, and analytical reagent. It also acts as a plating inhibitor on primary water piping.

Solubility
Soluble in water, alcohol, dilute mineral acids and alkalies.

Notes
Incompatible with zinc salts, alkalies and their carbonates, oxalates, phosphates, and sulfides. Hygroscopic in nature.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text