3-Bromo-4-hydroxypyridine, 97%, Thermo Scientific Chemicals
3-Bromo-4-hydroxypyridine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Bromo-4-hydroxypyridine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H33754.06
also known as H33754-06
Price (USD)/ Each
800.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
800.00
Add to cart
3-Bromo-4-hydroxypyridine, 97%, Thermo Scientific Chemicals
Catalog numberH33754.06
Price (USD)/ Each
800.00
-
Add to cart
Chemical Identifiers
CAS36953-41-0
IUPAC Name3-bromo-1,4-dihydropyridin-4-one
Molecular FormulaC5H4BrNO
InChI KeyJWHZKNRXTDYNJO-UHFFFAOYSA-N
SMILESBrC1=CNC=CC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Melting Point (clear melt)228.0-237.0?C
Assay (Silylated GC)≥96.0%
Appearance (Color)Cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only