Ethyl 3-cyclopropyl-3-oxopropionate, 96%, Thermo Scientific Chemicals
Ethyl 3-cyclopropyl-3-oxopropionate, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 3-cyclopropyl-3-oxopropionate, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H33756.06
also known as H33756-06
Price (USD)/ Each
220.65
Online exclusive
245.00 
Save 24.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
220.65
Online exclusive
245.00 
Save 24.35 (10%)
Add to cart
Ethyl 3-cyclopropyl-3-oxopropionate, 96%, Thermo Scientific Chemicals
Catalog numberH33756.06
Price (USD)/ Each
220.65
Online exclusive
245.00 
Save 24.35 (10%)
-
Add to cart
Chemical Identifiers
CAS24922-02-9
IUPAC Nameethyl 3-cyclopropyl-3-oxopropanoate
Molecular FormulaC8H12O3
InChI KeyLFSVADABIDBSBV-UHFFFAOYSA-N
SMILESCCOC(=O)CC(=O)C1CC1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥95.0%
Refractive Index1.4460-1.4520 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only