Thermo Scientific Chemicals

2,5-Difluoro-4-nitrobenzoic acid, 97%, Thermo Scientific Chemicals

Catalog number: H33757.06
5 g, Each
Thermo Scientific Chemicals

2,5-Difluoro-4-nitrobenzoic acid, 97%, Thermo Scientific Chemicals

Catalog number: H33757.06
5 g, Each
Quantity
Catalog number: H33757.06
also known as H33757-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
116465-48-6
IUPAC Name
2,5-difluoro-4-nitrobenzoic acid
Molecular Formula
C7H3F2NO4
InChI Key
GPTNSBLYGCZJQV-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC(F)=C(C=C1F)[N+]([O-])=O
Form
Powder
Melting Point (clear melt)
139.0-149.0?C
Assay (Silylated GC)
≥96.0%
Appearance (Color)
Pale cream
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%

Description

2,5-Difluoro-4-nitrobenzoic acid is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Difluoro-4-nitrobenzoic acid is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text