Thermo Scientific Chemicals

4-Chloro-5,6-dimethylthieno[2,3-d]pyrimidine, 96%, Thermo Scientific Chemicals

Catalog number: H33770.03
1 g, Each
Thermo Scientific Chemicals

4-Chloro-5,6-dimethylthieno[2,3-d]pyrimidine, 96%, Thermo Scientific Chemicals

Catalog number: H33770.03
1 g, Each
Quantity
Catalog number: H33770.03
also known as H33770-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
108831-68-1
Appearance (Color)
Pale brown
Form
Powder
Assay (HPLC)
≥95.0%
Sulfated Ash
≤1.0%

Description

Employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed containers. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text