Thermo Scientific Chemicals

12-Azidododecylphosphonic acid, 95%, Thermo Scientific Chemicals

Catalog number: H33784.MD
250 mg, Each
Thermo Scientific Chemicals

12-Azidododecylphosphonic acid, 95%, Thermo Scientific Chemicals

Catalog number: H33784.MD
250 mg, Each
Quantity
Catalog number: H33784.MD
also known as H33784-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
721457-32-5
Appearance (Color)
White
Assay (Aqueous acid-base Titration)
≥94.0 to ≤106.0%
Form
Crystals or powder or crystalline powder or flakes
Melting Point (clear melt)
83.5-92.5°C

Description

This coupling agent specifically reacts with alkyne functions to form the corresponding 1,2,3-triazole via a copper-free 1,3-dipolar cycloaddition.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
This coupling agent specifically reacts with alkyne functions to form the corresponding 1,2,3-triazole via a copper-free 1,3-dipolar cycloaddition.

Notes
Store in refrigerator. Protect from heat. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text