Thermo Scientific Chemicals

3-(1-Naphthyl)propionic acid, 96%, Thermo Scientific Chemicals

Catalog number: H33795.06
5 g, Each
Thermo Scientific Chemicals

3-(1-Naphthyl)propionic acid, 96%, Thermo Scientific Chemicals

Catalog number: H33795.06
5 g, Each
Quantity
Catalog number: H33795.06
also known as H33795-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3243-42-3
IUPAC Name
3-(naphthalen-1-yl)propanoic acid
Molecular Formula
C13H12O2
InChI Key
PRLKVVMRQFFIOQ-UHFFFAOYSA-N
SMILES
OC(=O)CCC1=C2C=CC=CC2=CC=C1
Appearance (Color)
White to pale yellow
Assay (Aqueous acid-base Titration)
≥95.0 to ≤105.0%
Melting Point (clear melt)
151.0-160.0?C
Form
Powder
Assay (Silylated GC)
≥95.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text