A phosphorous halide used for biochemical research. Also used as an intermediate in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
A phosphorous halide used for biochemical research. Also used as an intermediate in organic synthesis.
Solubility
Soluble in alcohol and chloroform. Slightly soluble in water.
Notes
Moisture Sensitive. Store away from water/moisture. Store away from oxidizing agent.
RUO – Research Use Only
General References:
- James McNulty; Venkatesan Krishnamoorthy; Al Robertson. Direct formation of esters and amides from carboxylic acids using diethyl chlorophosphate in pyridine. Tetrahedron Letters. 200849 (44), 6344-6347
- Zarei, Maaroof. One-pot sequence synthesis of azetidin-2-one using diethyl chlorophosphate. Journal of Chemical Research. 201236 (2), 118-120(3)