Thermo Scientific Chemicals

Ethyl 2-amino-5-phenylthiophene-3-carboxylate, 97%, Thermo Scientific Chemicals

Catalog number: H33823.06
5 g, Each
Thermo Scientific Chemicals

Ethyl 2-amino-5-phenylthiophene-3-carboxylate, 97%, Thermo Scientific Chemicals

Catalog number: H33823.06
5 g, Each
Quantity
Catalog number: H33823.06
also known as H33823-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4815-34-3
IUPAC Name
ethyl 2-amino-5-phenylthiophene-3-carboxylate
Molecular Formula
C13H13NO2S
InChI Key
WIVNPGXPJBBZQH-UHFFFAOYSA-N
SMILES
CCOC(=O)C1=C(N)SC(=C1)C1=CC=CC=C1
Melting Point (clear melt)
119.0-128.0?C
Appearance (Color)
Brown
Assay (GC)
≥96.0%
Form
Powder

Description

Ethyl 2-amino-5-phenylthiophene-3-carboxylate is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 2-amino-5-phenylthiophene-3-carboxylate is used as a pharmaceutical intermediate.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text