Thermo Scientific Chemicals

11-Aminoundecyltriethoxysilane, 90+%, Thermo Scientific Chemicals

Catalog number: H33835.MD
250 mg, Each
Thermo Scientific Chemicals

11-Aminoundecyltriethoxysilane, 90+%, Thermo Scientific Chemicals

Catalog number: H33835.MD
250 mg, Each
Quantity
Catalog number: H33835.MD
also known as H33835-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
116821-45-5
IUPAC Name
(11-aminoundecyl)triethoxysilane
Molecular Formula
C17H39NO3Si
InChI Key
LPWZCJFZJCOBHO-UHFFFAOYSA-N
SMILES
CCO[Si](CCCCCCCCCCCN)(OCC)OCC
Appearance (Color)
Clear colorless
Assay (GC)
≥90.0%
Refractive Index
1.4340-1.4420 @ 20?C
Identification (FTIR)
Conforms
Form
Liquid

Description

Used as primary and secondary intermediate for pharmaceutical and chemical additives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as primary and secondary intermediate for pharmaceutical and chemical additives.

Notes
Moisture Sensitive. Store away from water/moisture. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text