3-Chloro-2-hydrazino-5-(trifluoromethyl)pyridine, 97%, Thermo Scientific Chemicals
3-Chloro-2-hydrazino-5-(trifluoromethyl)pyridine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Chloro-2-hydrazino-5-(trifluoromethyl)pyridine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H33850.03
also known as H33850-03
Price (USD)/ Each
79.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
79.00
Add to cart
3-Chloro-2-hydrazino-5-(trifluoromethyl)pyridine, 97%, Thermo Scientific Chemicals
Catalog numberH33850.03
Price (USD)/ Each
79.00
-
Add to cart
Chemical Identifiers
CAS89570-82-1
IUPAC Name3-chloro-2-hydrazinyl-5-(trifluoromethyl)pyridine
Molecular FormulaC6H5ClF3N3
InChI KeyKHFKSHYCVQZAQP-UHFFFAOYSA-N
SMILESNNC1=NC=C(C=C1Cl)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream
Assay (GC)≥96.0%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)87.0-96.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only