Thermo Scientific Chemicals

6-Bromopyrazolo[1,5-a]pyrimidine, 97%, Thermo Scientific Chemicals

Catalog number: H33855.03
1 g, Each
Thermo Scientific Chemicals

6-Bromopyrazolo[1,5-a]pyrimidine, 97%, Thermo Scientific Chemicals

Catalog number: H33855.03
1 g, Each
Quantity
Catalog number: H33855.03
also known as H33855-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
705263-10-1
IUPAC Name
6-bromopyrazolo[1,5-a]pyrimidine
Molecular Formula
C6H4BrN3
InChI Key
VDHTXLUCUNPVLO-UHFFFAOYSA-N
SMILES
BrC1=CN2N=CC=C2N=C1
Form
Crystals or powder or crystalline powder or flakes
Melting Point (clear melt)
119.0-128.0?C
Assay (GC)
≥96.0%
Appearance (Color)
Pale yellow to yellow to pale orange

Description

6-Bromopyrazolo[1,5-a]pyrimidine is a reactant in the synthesis of LDN-212854, a selective and potent inhibitor of the BMP type I receptor kinases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Bromopyrazolo[1,5-a]pyrimidine is a reactant in the synthesis of LDN-212854, a selective and potent inhibitor of the BMP type I receptor kinases.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Store in dark. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text