3-Amino-4-hydroxybenzonitrile, 97%, Thermo Scientific Chemicals
3-Amino-4-hydroxybenzonitrile, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Amino-4-hydroxybenzonitrile, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H33856.06
also known as H33856-06
Price (USD)/ Each
647.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
647.00
Add to cart
3-Amino-4-hydroxybenzonitrile, 97%, Thermo Scientific Chemicals
Catalog numberH33856.06
Price (USD)/ Each
647.00
-
Add to cart
Chemical Identifiers
CAS14543-43-2
IUPAC Name3-amino-4-hydroxybenzonitrile
Molecular FormulaC7H6N2O
InChI KeyZEWCASRNRWXXSO-UHFFFAOYSA-N
SMILESNC1=CC(=CC=C1O)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Brown
Assay (GC)≥96.0%
Melting Point (clear melt)151.0-161.0?C
FormPowder
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only