Thermo Scientific Chemicals

Isopropylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog number: H33861.06
5 g, Each
Thermo Scientific Chemicals

Isopropylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog number: H33861.06
5 g, Each
Quantity
Catalog number: H33861.06
also known as H33861-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
76347-13-2
IUPAC Name
4,4,5,5-tetramethyl-2-(propan-2-yl)-1,3,2-dioxaborolane
Molecular Formula
C9H19BO2
InChI Key
MECCSFDHAVAAAW-UHFFFAOYSA-N
SMILES
CC(C)B1OC(C)(C)C(C)(C)O1
Form
Liquid
Refractive Index
1.4070-1.4110 @ 20°C
Appearance (Color)
Clear colorless
Assay (GC)
≥96.0%

Description

Isopropylboronic acid pinacol ester is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isopropylboronic acid pinacol ester is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text