2-Chloro-5-(trifluoromethyl)pyrazine, 97%, Thermo Scientific Chemicals
2-Chloro-5-(trifluoromethyl)pyrazine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Chloro-5-(trifluoromethyl)pyrazine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Catalog number H33863.03
also known as H33863-03
Price (USD)/ Each
185.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
185.00
Add to cart
2-Chloro-5-(trifluoromethyl)pyrazine, 97%, Thermo Scientific Chemicals
Catalog numberH33863.03
Price (USD)/ Each
185.00
-
Add to cart
Chemical Identifiers
CAS799557-87-2
IUPAC Name2-chloro-5-(trifluoromethyl)pyrazine
Molecular FormulaC5H2ClF3N2
InChI KeyAIEGIFIEQXZBCP-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CN=C(Cl)C=N1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.4440-1.4490 @ 20?C
Identification (FTIR)Conforms
Appearance (Color)Clear colorless
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only