Thermo Scientific Chemicals

3-[3-(Benzyloxy)phenyl]propionic acid, 96%, Thermo Scientific Chemicals

Catalog number: H33899.03
1 g, Each
Thermo Scientific Chemicals

3-[3-(Benzyloxy)phenyl]propionic acid, 96%, Thermo Scientific Chemicals

Catalog number: H33899.03
1 g, Each
Quantity
Catalog number: H33899.03
also known as H33899-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
57668-34-5
IUPAC Name
3-[3-(benzyloxy)phenyl]propanoic acid
Molecular Formula
C16H16O3
InChI Key
HVXOYUOKPLHWNU-UHFFFAOYSA-N
SMILES
OC(=O)CCC1=CC(OCC2=CC=CC=C2)=CC=C1
Appearance (Color)
White
Assay (Silylated GC)
≥95.0%
Assay (Aqueous acid-base Titration)
≥95.0 to ≤105.0%
Melting Point (clear melt)
79.5-88.5?C
Form
Crystals or powder or crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text