Thermo Scientific Chemicals

Methyl 4-bromopyrrole-2-carboxylate, 97%, Thermo Scientific Chemicals

Catalog number: H33918.06
5 g, Each
Thermo Scientific Chemicals

Methyl 4-bromopyrrole-2-carboxylate, 97%, Thermo Scientific Chemicals

Catalog number: H33918.06
5 g, Each
Quantity
Catalog number: H33918.06
also known as H33918-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
934-05-4
IUPAC Name
methyl 4-bromo-1H-pyrrole-2-carboxylate
Molecular Formula
C6H6BrNO2
InChI Key
ZXFCRVGOHJHZNF-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC(Br)=CN1
Appearance (Color)
White to cream or pale yellow
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
98.5-104.5?C
Identification (FTIR)
Conforms
Assay (GC)
≥96.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text