Diethyl 12-mercaptododecylphosphonate, 95%, Thermo Scientific Chemicals
Diethyl 12-mercaptododecylphosphonate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diethyl 12-mercaptododecylphosphonate, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H33935.MD
also known as H33935-MD
Price (USD)/ Each
151.00
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
151.00
Add to cart
Diethyl 12-mercaptododecylphosphonate, 95%, Thermo Scientific Chemicals
Catalog numberH33935.MD
Price (USD)/ Each
151.00
-
Add to cart
Chemical Identifiers
CAS1049677-30-6
IUPAC Namediethyl (12-sulfanyldodecyl)phosphonate
Molecular FormulaC16H35O3PS
InChI KeyJAVFXEXYSKLPHD-UHFFFAOYSA-N
SMILESCCOP(=O)(CCCCCCCCCCCCS)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥94.0%
FormLiquid
Refractive Index1.4640-1.4700 @ 20°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only