Thermo Scientific Chemicals

3-Methoxythiophene-2-carboxaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: H33940.MD
250 mg, Each
Thermo Scientific Chemicals

3-Methoxythiophene-2-carboxaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: H33940.MD
250 mg, Each
Quantity
Catalog number: H33940.MD
also known as H33940-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
35134-07-7
IUPAC Name
3-methoxythiophene-2-carbaldehyde
Molecular Formula
C6H6O2S
InChI Key
KGJDTMQUUPIAEF-UHFFFAOYSA-N
SMILES
COC1=C(SC=C1)C=O
Appearance (Color)
White to pale cream
Free acid (titration)
≤1.5%
Form
Crystals or powder or crystalline powder
Assay (GC)
≥96.0%
Melting Point (clear melt)
76.0-85.0?C

Description

3-Methoxythiophene-2-carboxaldehyde is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methoxythiophene-2-carboxaldehyde is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Store away from air, oxidizing agents. Store under inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text