Thermo Scientific Chemicals

Trimethylolpropane tris[3-(2-methylaziridin-1-yl)propionate], Thermo Scientific Chemicals

Catalog number: H33949.03
1 g, Each
Thermo Scientific Chemicals

Trimethylolpropane tris[3-(2-methylaziridin-1-yl)propionate], Thermo Scientific Chemicals

Catalog number: H33949.03
1 g, Each
Quantity
Catalog number: H33949.03
also known as H33949-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
64265-57-2
IUPAC Name
2,2-bis({[3-(2-methylaziridin-1-yl)propanoyl]oxy}methyl)butyl 3-(2-methylaziridin-1-yl)propanoate
Molecular Formula
C24H41N3O6
InChI Key
YKEGOEUSKXVSPN-UHFFFAOYNA-N
SMILES
CCC(COC(=O)CCN1CC1C)(COC(=O)CCN1CC1C)COC(=O)CCN1CC1C
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Assay (Non-aqueous acid-base Titration)
≥88.0%
Identification (FTIR)
Conforms
Refractive Index
1.4710-1.4750 @ 20°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text