Thermo Scientific Chemicals

4-Methylbenzoic anhydride, 97%, Thermo Scientific Chemicals

Catalog number: H33951.06
5 g, Each
Thermo Scientific Chemicals

4-Methylbenzoic anhydride, 97%, Thermo Scientific Chemicals

Catalog number: H33951.06
5 g, Each
Quantity
Catalog number: H33951.06
also known as H33951-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13222-85-0
IUPAC Name
4-methylbenzoyl 4-methylbenzoate
Molecular Formula
C16H14O3
InChI Key
BJMLLSSSTGHJJE-UHFFFAOYSA-N
SMILES
CC1=CC=C(C=C1)C(=O)OC(=O)C1=CC=C(C)C=C1
Melting Point (clear melt)
91.0-100.0?C
Form
Crystals or powder or crystalline powder
Appearance (Color)
White
Assay (GC)
≥96.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text