Thermo Scientific Chemicals

12-Mercaptododecylphosphonic acid, 95%, Thermo Scientific Chemicals

Catalog number: H33969.MD
250 mg, Each
Thermo Scientific Chemicals

12-Mercaptododecylphosphonic acid, 95%, Thermo Scientific Chemicals

Catalog number: H33969.MD
250 mg, Each
Quantity
Catalog number: H33969.MD
also known as H33969-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
159239-33-5
IUPAC Name
(12-sulfanyldodecyl)phosphonic acid
Molecular Formula
C12H27O3PS
InChI Key
PVIUMTORLYKRJT-UHFFFAOYSA-N
SMILES
OP(O)(=O)CCCCCCCCCCCCS
Assay (Aqueous acid-base Titration)
≥94.0 to ≤106%
Form
Crystals or powder or crystalline powder
Loss on Drying
≤1.5% (50?C, constant weight)
Appearance (Color)
White to cream
Melting Point
91-97?C

Description

Application to the Extraction of Heavy Metals. Recent works using phosphonic acid PA) molecules have demonstrated the ability of 12-mercaptododecylphosphonic acid (MDPA)to strengthen the bond between Au and TiO 2 , leading to an increase in h K.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Application to the Extraction of Heavy Metals. Recent works using phosphonic acid PA) molecules have demonstrated the ability of 12-mercaptododecylphosphonic acid (MDPA)to strengthen the bond between Au and TiO 2 , leading to an increase in h K.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text