3-(4,4-Dimethyl-2-oxazolinyl)pyridine, 97%, Thermo Scientific Chemicals
3-(4,4-Dimethyl-2-oxazolinyl)pyridine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-(4,4-Dimethyl-2-oxazolinyl)pyridine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
1 g
Catalog number H33997.09
also known as H33997-09
Price (USD)/ Each
210.00
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
210.00
Add to cart
3-(4,4-Dimethyl-2-oxazolinyl)pyridine, 97%, Thermo Scientific Chemicals
Catalog numberH33997.09
Price (USD)/ Each
210.00
-
Add to cart
Chemical Identifiers
CAS68981-86-2
IUPAC Name3-(4,4-dimethyl-4,5-dihydro-1,3-oxazol-2-yl)pyridine
Molecular FormulaC10H12N2O
InChI KeyXYABHURTMBEOGW-UHFFFAOYSA-N
SMILESCC1(C)COC(=N1)C1=CN=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Refractive Index1.5295-1.5345 @ 20?C
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only