Methyl 3-aminopyridine-4-carboxylate, 97%, Thermo Scientific Chemicals
Methyl 3-aminopyridine-4-carboxylate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl 3-aminopyridine-4-carboxylate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
1 g
Catalog number H34007.09
also known as H34007-09
Price (USD)/ Each
474.00
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
474.00
Add to cart
Methyl 3-aminopyridine-4-carboxylate, 97%, Thermo Scientific Chemicals
Catalog numberH34007.09
Price (USD)/ Each
474.00
-
Add to cart
Chemical Identifiers
CAS55279-30-6
IUPAC Namemethyl 3-aminopyridine-4-carboxylate
Molecular FormulaC7H8N2O2
InChI KeyXLQIGLBALJNHKR-UHFFFAOYSA-N
SMILESCOC(=O)C1=C(N)C=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to yellow
FormPowder
Assay (GC)≥96.0%
Melting Point (clear melt)79.0-88.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only