2,4-Dichloropyridine-3-carboxaldehyde, 97%, Thermo Scientific Chemicals
2,4-Dichloropyridine-3-carboxaldehyde, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,4-Dichloropyridine-3-carboxaldehyde, 97%, Thermo Scientific Chemicals

Have Questions?
Quantity:
1 g
Catalog number H34053.03
also known as H34053-03
Price (USD)/ Each
153.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
153.00
Add to cart
2,4-Dichloropyridine-3-carboxaldehyde, 97%, Thermo Scientific Chemicals
Catalog numberH34053.03
Price (USD)/ Each
153.00
-
Add to cart
Chemical Identifiers
CAS134031-24-6
IUPAC Name2,4-dichloropyridine-3-carbaldehyde
Molecular FormulaC6H3Cl2NO
InChI KeyGWBHJHIZHNTJLA-UHFFFAOYSA-N
SMILESClC1=C(C=O)C(Cl)=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Assay (GC)≥96.0%
Free acid (titration)≤1.5%
Melting Point (clear melt)67.0-77.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only