8-Methoxyjulolidine, 95%, Thermo Scientific Chemicals
8-Methoxyjulolidine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

8-Methoxyjulolidine, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
Catalog number H34100.03
also known as H34100-03
Price (USD)/ Each
46.70
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
46.70
Add to cart
8-Methoxyjulolidine, 95%, Thermo Scientific Chemicals
Catalog numberH34100.03
Price (USD)/ Each
46.70
-
Add to cart
Chemical Identifiers
CAS63468-83-7
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥94.0%
Refractive Index1.5790-1.5850 @ 20?C
Appearance (Color)Clear red to brown
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only