Thermo Scientific Chemicals

Ethyl 2-amino-5,6,7,8-tetrahydro-4H-cyclohepta[b]thiophene-3-carboxylate, 96%, Thermo Scientific Chemicals

Catalog number: H34116.03
1 g, Each
Thermo Scientific Chemicals

Ethyl 2-amino-5,6,7,8-tetrahydro-4H-cyclohepta[b]thiophene-3-carboxylate, 96%, Thermo Scientific Chemicals

Catalog number: H34116.03
1 g, Each
Quantity
Catalog number: H34116.03
also known as H34116-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
40106-13-6
IUPAC Name
ethyl 2-amino-4H,5H,6H,7H,8H-cyclohepta[b]thiophene-3-carboxylate
Molecular Formula
C12H17NO2S
InChI Key
XTUHIGALMIGZST-UHFFFAOYSA-N
SMILES
CCOC(=O)C1=C(N)SC2=C1CCCCC2
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
85-91?C
Proton NMR
Conforms to structure
Assay (GC)
≥95.0%
Appearance (Color)
White to pale cream or pale yellow to pale orange

Description

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Soluble in methanol and ethanol.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text