5-(2-Pyridyl)-1,3-oxazole, 97%, Thermo Scientific Chemicals
5-(2-Pyridyl)-1,3-oxazole, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-(2-Pyridyl)-1,3-oxazole, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 mg
1 g
Catalog number H34123.06
also known as H34123-06
Price (USD)/ Each
312.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
312.00
Add to cart
5-(2-Pyridyl)-1,3-oxazole, 97%, Thermo Scientific Chemicals
Catalog numberH34123.06
Price (USD)/ Each
312.00
-
Add to cart
Chemical Identifiers
CAS70380-73-3
IUPAC Name2-(1,3-oxazol-5-yl)pyridine
Molecular FormulaC8H6N2O
InChI KeyLUMKXMDGONYZHO-UHFFFAOYSA-N
SMILESO1C=NC=C1C1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.5955-1.6005 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only