1-Methylindole-3-carbonitrile, 96%, Thermo Scientific Chemicals
1-Methylindole-3-carbonitrile, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Methylindole-3-carbonitrile, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
5 g
Catalog number H34157.03
also known as H34157-03
Price (USD)/ Each
99.20
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
99.20
Add to cart
1-Methylindole-3-carbonitrile, 96%, Thermo Scientific Chemicals
Catalog numberH34157.03
Price (USD)/ Each
99.20
-
Add to cart
Chemical Identifiers
CAS24662-37-1
IUPAC Name1-methyl-1H-indole-3-carbonitrile
Molecular FormulaC10H8N2
InChI KeyFBAXZPMXGBNBPE-UHFFFAOYSA-N
SMILESCN1C=C(C#N)C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale orange
FormPowder
Assay (GC)≥95.0%
Melting Point (clear melt)51.5-60.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only