2-Bromo-2'-fluoroacetophenone, 98%, Thermo Scientific Chemicals
2-Bromo-2'-fluoroacetophenone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Bromo-2'-fluoroacetophenone, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
1 g
Catalog number H34179.09
also known as H34179-09
Price (USD)/ Each
223.00
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
223.00
Add to cart
2-Bromo-2'-fluoroacetophenone, 98%, Thermo Scientific Chemicals
Catalog numberH34179.09
Price (USD)/ Each
223.00
-
Add to cart
Chemical Identifiers
CAS655-15-2
IUPAC Name2-bromo-1-(2-fluorophenyl)ethan-1-one
Molecular FormulaC8H6BrFO
InChI KeyQDNWNJSLWKHNTM-UHFFFAOYSA-N
SMILESFC1=CC=CC=C1C(=O)CBr
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)27-34°C
FormCrystals or powder or crystalline powder or lumpy powder or fused solid
Appearance (Color)Pale cream to cream to brown to pale gray or pale blue
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only