3-Bromo-4-iodopyridine, 97%, Thermo Scientific Chemicals
3-Bromo-4-iodopyridine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Bromo-4-iodopyridine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H34203.MD
also known as H34203-MD
Price (USD)/ Each
53.10
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
53.10
Add to cart
3-Bromo-4-iodopyridine, 97%, Thermo Scientific Chemicals
Catalog numberH34203.MD
Price (USD)/ Each
53.10
-
Add to cart
Chemical Identifiers
CAS89167-19-1
IUPAC Name3-bromo-4-iodopyridine
Molecular FormulaC5H3BrIN
InChI KeyLKDNPXCBLVOBRP-UHFFFAOYSA-N
SMILESBrC1=C(I)C=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pink
Assay (GC)≥96.0%
Melting Point (clear melt)109.0-118.0?C
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only