Thermo Scientific Chemicals

3,3'-Di-n-propylthiadicarbocyanine iodide, 96%, Thermo Scientific Chemicals

Catalog number: H34209.03
1 g, Each
Thermo Scientific Chemicals

3,3'-Di-n-propylthiadicarbocyanine iodide, 96%, Thermo Scientific Chemicals

Catalog number: H34209.03
1 g, Each
Quantity
Catalog number: H34209.03
also known as H34209-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
53213-94-8
IUPAC Name
3-propyl-2-[(3E)-5-[(2Z)-3-propyl-2,3-dihydro-1,3-benzothiazol-2-ylidene]penta-1,3-dien-1-yl]-1,3-benzothiazol-3-ium iodide
Molecular Formula
C25H27IN2S2
InChI Key
GDEURKKLNUGTDA-UHFFFAOYSA-M
SMILES
[I-].CCCN1C(SC2=CC=CC=C12)=CC=CC=CC1=[N+](CCC)C2=CC=CC=C2S1
Elemental Analysis
N: 4.87-5.38% (Theory: 5.13%)
Elemental Analysis
S: 11.15-12.32% (Theory: 11.73%)
lambda-max (UV-Vis)
645-660nm
Appearance (Color)
Pale yellow to yellow to green to dark green
Form
Crystals or powder or crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text